This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02307200_m1
See other CAL GE Assays ›
Gene Symbol:
CAL
Gene Name:
K-box region and MADS-box transcription factor family protein
Gene Aliases:
AGAMOUS-like 10, AGL10, AT1G26310, CAL1, CAULIFLOWER, F28B23.25, F28B23_25, MADS3
Chromosome Location:
Chr.1: 9099994 - 9103644 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02307200_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_102395.3 NP_564243.1 6 - 7 645 84
GenBank mRNA AB493477.1 - 6 - 7 511 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
positive regulation of flower development
floral meristem determinacy
positive regulation of transcription from RNA polymerase II promoter
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0