This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02304130_g1
See other VSP1 GE Assays ›
Gene Symbol:
VSP1
Gene Name:
vegetative storage protein 1
Gene Aliases:
AT5G24780, ATVSP1, T4C12.50, VEGETATIVE STORAGE PROTEIN 1, vegetative storage protein 1
Chromosome Location:
Chr.5: 8507590 - 8509201 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02304130_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122387.4 NP_568455.1 2 - 3 953 84
GenBank mRNA AF386930.1 - 2 - 3 679 84
AY044328.1 - 2 - 3 643 84
AY072506.1 - 2 - 3 641 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

defense response
response to jasmonic acid
dephosphorylation
acid phosphatase activity
transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0