This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02304128_g1
See other VSP2 GE Assays ›
Gene Symbol:
VSP2
Gene Name:
vegetative storage protein 2
Gene Aliases:
AT5G24770, ATVSP2, T4C12.40, VEGETATIVE STORAGE PROTEIN 2, vegetative storage protein 2
Chromosome Location:
Chr.5: 8500468 - 8502224 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02304128_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122386.4 NP_568454.1 2 - 3 999 69
GenBank mRNA AY048282.1 - 2 - 3 655 69
AY050819.1 - 2 - 3 655 69
AY092991.1 - 2 - 3 633 69
AY114083.1 - 2 - 3 619 69
AY114606.1 - 2 - 3 619 69
BT000761.1 - 2 - 3 659 69
BT000762.1 - 2 - 3 657 69
BT006341.1 - 2 - 3 619 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

defense response to insect
response to oxidative stress
response to wounding
response to insect
response to jasmonic acid
hyperosmotic salinity response
response to copper ion
acid phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0