This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02304020_g1
See other AT5G24650 GE Assays ›
Gene Symbol:
AT5G24650
Gene Name:
Mitochondrial import inner membrane translocase subunit Tim17/Tim22/Tim23 family protein
Gene Aliases:
AT5G24650, HP30-2, K18P6.19, K18P6_19, hypothetical protein 30-2
Chromosome Location:
Chr.5: 8437024 - 8439104 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02304020_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G24650
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122373.3 NP_197853.1 3 - 4 628 90
GenBank mRNA AY070735.1 - 3 - 4 567 90
AY142048.1 - 3 - 4 529 90
DQ405267.1 - 3 - 4 529 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

mitochondrion organization
tRNA import into mitochondrion
protein targeting to chloroplast
protein import into mitochondrial inner membrane
protein binding
protein transmembrane transporter activity
protein self-association

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0