This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02303872_g1
See other AT5G24350 GE Assays ›
Gene Symbol:
AT5G24350
Gene Name:
neuroblastoma-amplified sequence protein
Gene Aliases:
AT5G24350, K16H17.4, K16H17_4, MAG2-interacting protein 2, MIP2
Chromosome Location:
Chr.5: 8301105 - 8310895 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02303872_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G24350
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203451.1 NP_001190380.1 12 - 13 4559 146
NM_001343876.1 NP_001318636.1 11 - 12 4514 146
NM_122343.3 NP_197823.1 11 - 12 4499 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

vacuolar protein processing
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport
protein exit from endoplasmic reticulum
protein maturation
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0