This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02303432_g1
See other PDR2 GE Assays ›
Gene Symbol:
PDR2
Gene Name:
phosphate deficiency response 2
Gene Aliases:
AT5G23630, MALE GAMETOGENESIS IMPAIRED ANTHERS, MIA, MQM1.11, MQM1_11, PI DEFICIENCY RESPONSE 2, phosphate deficiency response 2
Chromosome Location:
Chr.5: 7960478 - 7968197 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02303432_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122268.5 NP_197752.1 9 - 10 1592 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cation transport
cellular calcium ion homeostasis
cellular metal ion homeostasis
pollen germination
meristem maintenance
pollen maturation
cellular response to phosphate starvation
stem cell fate determination
ATPase activity
cation-transporting ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0