This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02302942_g1
See other MGT10 GE Assays ›
Gene Symbol:
MGT10
Gene Name:
magnesium (Mg) transporter 10
Gene Aliases:
AT5G22830, ATMGT10, GMN10, MAGNESIUM TRANSPORTER 10, MRN17.6, MRN17_6, MRS2-11, magnesium (Mg) transporter 10
Chromosome Location:
Chr.5: 7627597 - 7630993 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02302942_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGT10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001343773.1 NP_001332159.1 9 - 10 1157 91
NM_122188.3 NP_568424.1 9 - 10 1157 91
GenBank mRNA BT029184.1 - 9 - 10 1078 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

thylakoid membrane organization
photoprotection
magnesium ion homeostasis
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0