This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02302413_g1
See other B73 GE Assays ›
Gene Symbol:
B73
Gene Name:
molybdopterin biosynthesis CNX1 protein / molybdenum cofactor biosynthesis enzyme CNX1 (CNX1)
Gene Aliases:
AT5G20990, CHL6, CHLORATE RESISTANT 6, CNX, CNX1, CO-FACTOR FOR NITRATE REDUCTASE AND XANTHINE DEHYDROGENASE, CO-FACTOR FOR NITRATE REDUCTASE AND XANTHINE DEHYDROGENASE 1, MOLYBDENUM COFACTOR BIOSYNTHESIS ENZYME CNX1, SIR4, SIRTINOL 4, T10F18.20, T10F18_20
Chromosome Location:
Chr.5: 7128475 - 7133512 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02302413_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001343692.1 NP_001332397.1 6 - 7 1678 71
NM_001343693.1 NP_001332398.1 6 - 7 921 71
NM_122108.4 NP_197599.1 7 - 8 1181 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

Mo-molybdopterin cofactor biosynthetic process
auxin-activated signaling pathway
molybdenum incorporation into molybdenum-molybdopterin complex
molybdopterin cofactor biosynthetic process
molybdenum ion binding
molybdopterin adenylyltransferase activity
molybdopterin molybdotransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header