This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02296053_g1
See other APC8 GE Assays ›
Gene Symbol:
APC8
Gene Name:
anaphase-promoting complex subunit 8
Gene Aliases:
AT3G48150, CDC23, anaphase-promoting complex subunit 8
Chromosome Location:
Chr.3: 17779507 - 17782682 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02296053_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APC8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_114684.5 NP_190398.1 1 - 2 207 71
GenBank mRNA AY057562.1 - 1 - 2 170 71
AY099779.1 - 1 - 2 170 71
BT000289.1 - 1 - 2 90 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

metaphase/anaphase transition of mitotic cell cycle
protein ubiquitination
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
regulation of defense response
positive regulation of mitotic metaphase/anaphase transition
cell division
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0