This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02291835_g1
See other ASD1 GE Assays ›
Gene Symbol:
ASD1
Gene Name:
alpha-L-arabinofuranosidase 1
Gene Aliases:
ALPHA-L-ARABINOFURANOSIDASE, ALPHA-L-ARABINOFURANOSIDASE 1, ARABIDOPSIS THALIANA ALPHA-L-ARABINOFURANOSIDASE 1, ARAF, ARAF1, AT3G10740, ATASD1, alpha-L-arabinofuranosidase 1
Chromosome Location:
Chr.3: 3360711 - 3365739 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02291835_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337892.1 NP_001326565.1 11 - 12 1997 155
NM_001337893.1 NP_001326566.1 11 - 12 2146 155
NM_001337894.1 NP_001326567.1 10 - 11 1271 155
NM_111911.5 NP_187685.1 11 - 12 1809 155

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

xylan catabolic process
L-arabinose metabolic process
xylan 1,4-beta-xylosidase activity
alpha-L-arabinofuranosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0