This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02291700_gH
See other MTM1 GE Assays ›
Gene Symbol:
MTM1
Gene Name:
Myotubularin-like phosphatases II superfamily
Gene Aliases:
AT3G10550, Arabidopsis thaliana myotubularin 1, AtMTM1, myotubularin 1
Chromosome Location:
Chr.3: 3292890 - 3298621 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02291700_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111891.7 NP_187666.5 15 - 16 1800 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
intracellular signal transduction
cellular response to water deprivation
phosphatidylinositol dephosphorylation
regulation of response to water deprivation
phosphatidylinositol-3-phosphatase activity
protein tyrosine phosphatase activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0