This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02291072_g1
See other AT3G09720 GE Assays ›
Gene Symbol:
AT3G09720
Gene Name:
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases:
AT3G09720, AtRH57, RH57, RNA helicase 57
Chromosome Location:
Chr.3: 2979772 - 2983618 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02291072_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G09720
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337839.1 NP_001326599.1 3 - 4 406 92
NM_111806.7 NP_187583.2 3 - 4 377 92
GenBank mRNA AK227326.1 - 3 - 4 294 92
BT005833.1 - 2 - 3 216 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

maturation of SSU-rRNA
RNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0