This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02290106_gH
See other CCR2 GE Assays ›
Gene Symbol:
CCR2
Gene Name:
cinnamoyl coa reductase
Gene Aliases:
AT1G80820, ATCCR2, F23A5.17, F23A5_17, cinnamoyl coa reductase
Chromosome Location:
Chr.1: 30370474 - 30372799 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02290106_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335015.1 NP_001323345.1 2 - 3 629 72
NM_106730.4 NP_178197.1 2 - 3 629 72
GenBank mRNA AK227576.1 - 2 - 3 530 72
BT005826.1 - 2 - 3 457 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

circadian rhythm
response to cold
phenylpropanoid biosynthetic process
lignin biosynthetic process
negative regulation of circadian rhythm
catalytic activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
cinnamoyl-CoA reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0