This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02289442_g1
See other TED3 GE Assays ›
Gene Symbol:
TED3
Gene Name:
Pex2/Pex12 N-terminal domain-containing protein / zinc finger (C3HC4-type RING finger) family protein
Gene Aliases:
ARABIDOPSIS PEROXIN 2, AT1G79810, ATPEX2, F20B17.23, F20B17_23, PEROXIN 2, PEX2, PEX2P, REVERSAL OF THE DET PHENOTYPE 3
Chromosome Location:
Chr.1: 30019744 - 30022486 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02289442_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TED3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106630.3 NP_565222.1 6 - 7 924 60
NM_202454.2 NP_974183.1 6 - 7 928 60
GenBank mRNA AY064063.1 - 6 - 7 925 60
AY096373.1 - 6 - 7 724 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein monoubiquitination
fatty acid beta-oxidation
peroxisome organization
photomorphogenesis
protein import into peroxisome matrix
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0