This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02287695_g1
See other ADH1 GE Assays ›
Gene Symbol:
ADH1
Gene Name:
alcohol dehydrogenase 1
Gene Aliases:
ADH, ALCOHOL DEHYDROGENASE, ARABIDOPSIS THALIANA ALCOHOL DEHYDROGENASE, AT1G77120, ATADH, ATADH1, F22K20.19, F22K20_19, alcohol dehydrogenase 1
Chromosome Location:
Chr.1: 28975255 - 28977417 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02287695_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106362.3 NP_177837.1 4 - 5 959 82
GenBank mRNA AY045612.1 - 4 - 5 760 82
AY090330.1 - 4 - 5 705 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
response to osmotic stress
response to cold
response to flooding
response to water deprivation
response to salt stress
response to abscisic acid
response to sucrose
response to caffeine
response to estradiol
response to hydrogen peroxide
cellular respiration
formaldehyde catabolic process
response to cadmium ion
cellular response to hypoxia
positive regulation of cellular response to hypoxia
alcohol dehydrogenase (NAD) activity
alcohol dehydrogenase activity, zinc-dependent
zinc ion binding
protein homodimerization activity
S-(hydroxymethyl)glutathione dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0