This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02287188_g1
See other AT1G76350 GE Assays ›
Gene Symbol:
AT1G76350
Gene Name:
Plant regulator RWP-RK family protein
Gene Aliases:
AT1G76350, F15M4.15, F15M4_15, NIN-like protein 5, NLP5
Chromosome Location:
Chr.1: 28639453 - 28643086 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02287188_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G76350
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334724.1 NP_001321850.1 2 - 3 1199 69
NM_001334725.1 NP_001321849.1 3 - 4 1011 69
NM_001334726.1 NP_001321851.1 3 - 4 898 69
NM_106284.5 NP_177761.1 3 - 4 1183 69
GenBank mRNA AB493537.1 - 2 - 3 847 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header