This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02286785_g1
See other XRN3 GE Assays ›
Gene Symbol:
XRN3
Gene Name:
5'-3' exoribonuclease 3
Gene Aliases:
5'-3' EXORIBONUCLEASE 3, 5'-3' exoribonuclease 3, AT1G75660, AtXRN3, F10A5.15, F10A5_15
Chromosome Location:
Chr.1: 28408134 - 28415118 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02286785_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XRN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106217.3 NP_565114.1 21 - 22 2578 87
GenBank mRNA AF286719.1 - 21 - 22 2497 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process
rRNA 5'-end processing
miRNA catabolic process
negative regulation of posttranscriptional gene silencing
nuclear polyadenylation-dependent rRNA catabolic process
RNA binding
5'-3' exoribonuclease activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0