This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02286463_m1
See other AT1G75090 GE Assays ›
Gene Symbol:
AT1G75090
Gene Name:
DNA glycosylase superfamily protein
Gene Aliases:
AT1G75090, F9E10.6, F9E10_6
Chromosome Location:
Chr.1: 28187447 - 28189765 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02286463_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G75090
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106165.3 NP_565100.1 1 - 2 540 63
GenBank mRNA AY034999.1 - 1 - 2 500 63
AY063008.1 - 1 - 2 387 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
DNA-3-methyladenine glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0