This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02286431_g1
See other TIFY10B GE Assays ›
Gene Symbol:
TIFY10B
Gene Name:
TIFY domain/Divergent CCT motif family protein
Gene Aliases:
AT1G74950, F25A4.8, F25A4_8, JASMONATE-ZIM-DOMAIN PROTEIN 2, JAZ2
Chromosome Location:
Chr.1: 28148574 - 28150446 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02286431_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIFY10B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334673.1 NP_001322261.1 1 - 2 337 83
NM_106153.4 NP_565096.1 1 - 2 339 83
GenBank mRNA AF344319.1 - 1 - 2 151 83
AF386959.1 - 1 - 2 287 83
BT006301.1 - 1 - 2 151 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to wounding
response to jasmonic acid
regulation of defense response
regulation of jasmonic acid mediated signaling pathway
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0