This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02281256_g1
See other MOS14 GE Assays ›
Gene Symbol:
MOS14
Gene Name:
ARM repeat superfamily protein
Gene Aliases:
14, AT5G62600, K19B1.1, K19B1_1, modifier of snc1-1
Chromosome Location:
Chr.5: 25122803 - 25132716 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02281256_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOS14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125655.4 NP_201066.2 21 - 22 2244 75
GenBank mRNA AK229829.1 - 21 - 22 2170 75
AK230415.1 - 21 - 22 2205 75
BT002408.1 - 21 - 22 2194 75
BT010552.1 - 21 - 22 2001 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
regulation of RNA splicing
protein binding
Ran GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header