This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02281050_g1
See other ARP2 GE Assays ›
Gene Symbol:
ARP2
Gene Name:
actin related protein 2
Gene Aliases:
ACTIN RELATED PROTEIN 2, AT3G27000, ATARP2, WRM, WURM, actin related protein 2
Chromosome Location:
Chr.3: 9952479 - 9955982 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02281050_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113614.3 NP_189336.1 2 - 3 343 76
GenBank mRNA AF507910.1 - 2 - 3 145 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
actin filament organization
multidimensional cell growth
trichome morphogenesis
actin filament-based process
actin cytoskeleton organization
Arp2/3 complex-mediated actin nucleation
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0