This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02280153_g1
See other ATL5 GE Assays ›
Gene Symbol:
ATL5
Gene Name:
ribosomal protein L5
Gene Aliases:
AT3G25520, OLI5, OLIGOCELLULA 5, PGY3, PIGGYBACK3, RIBOSOMAL PROTEIN L5, RIBOSOMAL PROTEIN L5 A, RPL5A, ribosomal protein L5
Chromosome Location:
Chr.3: 9269277 - 9271465 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02280153_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATL5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203041.1 NP_001189970.1 4 - 5 296 75
NM_001338753.1 NP_001327862.1 5 - 6 741 75
NM_113448.5 NP_566767.1 5 - 6 621 75
GenBank mRNA AY054161.1 - 5 - 6 530 75
AY065103.1 - 5 - 6 540 75
AY081701.1 - 4 - 5 483 75
AY136319.1 - 5 - 6 542 75
AY186611.1 - 4 - 5 483 75
BT000411.1 - 4 - 5 483 75
BT002427.1 - 5 - 6 540 75
BT008705.1 - 4 - 5 483 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
translation
nucleocytoplasmic transport
adaxial/abaxial pattern specification
leaf morphogenesis
root morphogenesis
cell division
RNA binding
mRNA binding
structural constituent of ribosome
5S rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0