This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02278147_g1
See other AT3G22600 GE Assays ›
Gene Symbol:
AT3G22600
Gene Name:
Bifunctional inhibitor/lipid-transfer protein/seed storage 2S albumin superfamily protein
Gene Aliases:
AT3G22600, LTPG5, glycosylphosphatidylinositol-anchored lipid protein transfer 5
Chromosome Location:
Chr.3: 8006249 - 8007836 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02278147_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G22600
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113159.4 NP_566712.1 2 - 3 815 82
GenBank mRNA AB246327.1 - 2 - 3 376 82
AY045650.1 - 2 - 3 450 82
AY079122.1 - 2 - 3 376 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0