This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02277934_g1
See other POP2 GE Assays ›
Gene Symbol:
POP2
Gene Name:
Pyridoxal phosphate (PLP)-dependent transferases superfamily protein
Gene Aliases:
AT3G22200, GABA-T, GAMMA-AMINOBUTYRATE TRANSAMINASE, HER1, HEXENAL RESPONSE1, POLLEN-PISTIL INCOMPATIBILITY 2
Chromosome Location:
Chr.3: 7835144 - 7839061 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02277934_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203018.1 NP_001189947.1 3 - 4 569 97
NM_113117.4 NP_566700.1 3 - 4 366 97
GenBank mRNA AY034923.1 - 3 - 4 357 97
AY142571.1 - 3 - 4 229 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

sucrose metabolic process
inositol metabolic process
succinate metabolic process
glutamate metabolic process
glutamate decarboxylation to succinate
glutamine metabolic process
response to oxidative stress
biotin biosynthetic process
gamma-aminobutyric acid metabolic process
gamma-aminobutyric acid catabolic process
response to salt stress
pollen tube growth
pollen tube adhesion
adaxial/abaxial axis specification
response to organic substance
fruit development
pollen tube guidance
beta-alanine catabolic process
response to cadmium ion
root development
leaf development
shoot system development
4-aminobutyrate transaminase activity
adenosylmethionine-8-amino-7-oxononanoate transaminase activity
zinc ion binding
pyridoxal phosphate binding
4-aminobutyrate:pyruvate transaminase activity
cobalt ion binding
gamma-aminobutyrate transaminase (glyoxylate dependent) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0