This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02277414_g1
See other AT3G21300 GE Assays ›
Gene Symbol:
AT3G21300
Gene Name:
RNA methyltransferase family protein
Gene Aliases:
AT3G21300
Chromosome Location:
Chr.3: 7492739 - 7496371 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02277414_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G21300
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_113025.4 NP_188767.2 9 - 10 1666 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

response to cold
mRNA binding
tRNA (m5U54) methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0