This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02275353_m1
See other NPC2 GE Assays ›
Gene Symbol:
NPC2
Gene Name:
non-specific phospholipase C2
Gene Aliases:
AT2G26870, F12C20.9, F12C20_9, non-specific phospholipase C2
Chromosome Location:
Chr.2: 11456926 - 11459481 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02275353_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_128244.4 NP_180255.1 1 - 2 592 120
GenBank mRNA AB084292.1 - 1 - 2 466 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid catabolic process
gametophyte development
root development
phospholipase C activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0