This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02267944_g1
See other ADCL GE Assays ›
Gene Symbol:
ADCL
Gene Name:
D-aminoacid aminotransferase-like PLP-dependent enzymes superfamily protein
Gene Aliases:
4-amino-4-deoxychorismate lyase, AT5G57850, MTI20.10, MTI20_10
Chromosome Location:
Chr.5: 23435291 - 23437347 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02267944_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADCL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125170.5 NP_200593.2 1 - 2 189 67
GenBank mRNA AK226473.1 - 1 - 2 169 67
AY099783.1 - 1 - 2 168 67
AY128874.1 - 1 - 2 129 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

para-aminobenzoic acid biosynthetic process
branched-chain amino acid biosynthetic process
tetrahydrofolate biosynthetic process
branched-chain-amino-acid transaminase activity
4-amino-4-deoxychorismate lyase activity
D-alanine:2-oxoglutarate aminotransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0