This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02267889_g1
See other XBAT32 GE Assays ›
Gene Symbol:
XBAT32
Gene Name:
hypothetical protein
Gene Aliases:
AT5G57740, MRI1.10, MRI1_10, XB3 ortholog 2 in Arabidopsis thaliana
Chromosome Location:
Chr.5: 23394585 - 23397650 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02267889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XBAT32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125157.3 NP_200582.3 3 - 4 826 92
GenBank mRNA BT011688.1 - 3 - 4 698 92
BT012272.1 - 3 - 4 321 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
response to auxin
lateral root formation
negative regulation of ethylene biosynthetic process
protein ubiquitination
protein autoubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0