This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02266919_g1
See other MLO4 GE Assays ›
Gene Symbol:
MLO4
Gene Name:
Seven transmembrane MLO family protein
Gene Aliases:
AT1G11000, ATMLO4, MILDEW RESISTANCE LOCUS O 4, T19D16.26, T19D16_26
Chromosome Location:
Chr.1: 3671756 - 3676133 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02266919_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLO4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331947.1 NP_001320915.1 5 - 6 722 67
NM_001331948.1 NP_001320916.1 6 - 7 714 67
NM_100975.3 NP_563882.1 5 - 6 730 67
GenBank mRNA AF369565.1 - 5 - 6 637 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0