This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02266661_g1
See other AMSH2 GE Assays ›
Gene Symbol:
AMSH2
Gene Name:
associated molecule with the SH3 domain of STAM 2
Gene Aliases:
AT1G10600, F20B24.2, associated molecule with the SH3 domain of STAM 2
Chromosome Location:
Chr.1: 3503640 - 3506159 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02266661_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AMSH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084036.1 NP_001077505.1 3 - 4 367 147
NM_001331916.1 NP_001323286.1 3 - 4 431 147
NM_100936.3 NP_172530.2 3 - 4 395 147
GenBank mRNA AK175458.1 - 3 - 4 347 147
BT012645.1 - 3 - 4 306 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

protein deubiquitination
protein K63-linked deubiquitination
protein K48-linked deubiquitination
thiol-dependent ubiquitin-specific protease activity
isopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0