This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02266008_g1
See other IPGAM1 GE Assays ›
Gene Symbol:
iPGAM1
Gene Name:
Phosphoglycerate mutase, 2,3-bisphosphoglycerate-independent
Gene Aliases:
2, 3-biphosphoglycerate-independent phosphoglycerate mutase 1, AT1G09780, F21M12.16, F21M12_16
Chromosome Location:
Chr.1: 3165370 - 3167924 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02266008_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: iPGAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_100850.3 NP_563852.1 5 - 6 1463 61
GenBank mRNA AY086182.1 - 5 - 6 1408 61
AY150432.1 - 5 - 6 1351 61
BT000773.1 - 5 - 6 1408 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glucose catabolic process
glycolytic process
response to cold
pollen development
response to blue light
response to abscisic acid
response to carbon dioxide
stomatal movement
cellular carbohydrate metabolic process
response to cadmium ion
manganese ion binding
2,3-bisphosphoglycerate-independent phosphoglycerate mutase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0