This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02266005_g1
See other IPGAM1 GE Assays ›
Gene Symbol:
iPGAM1
Gene Name:
Phosphoglycerate mutase, 2,3-bisphosphoglycerate-independent
Gene Aliases:
2, 3-biphosphoglycerate-independent phosphoglycerate mutase 1, AT1G09780, F21M12.16, F21M12_16
Chromosome Location:
Chr.1: 3165370 - 3167924 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02266005_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: iPGAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_100850.3 NP_563852.1 2 - 3 545 73
GenBank mRNA AY086182.1 - 2 - 3 490 73
AY150432.1 - 2 - 3 433 73
BT000773.1 - 2 - 3 490 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glucose catabolic process
glycolytic process
response to cold
pollen development
response to blue light
response to abscisic acid
response to carbon dioxide
stomatal movement
cellular carbohydrate metabolic process
response to cadmium ion
manganese ion binding
2,3-bisphosphoglycerate-independent phosphoglycerate mutase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header