This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02264276_g1
See other UNE5 GE Assays ›
Gene Symbol:
UNE5
Gene Name:
thioredoxin family protein
Gene Aliases:
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 11, AT2G47470, ATPDI11, ATPDIL2-1, MATERNAL EFFECT EMBRYO ARREST 30, MEE30, PDI-LIKE 2-1, PDI11, PROTEIN DISULFIDE ISOMERASE 11, T30B22.23, UNFERTILIZED EMBRYO SAC 5
Chromosome Location:
Chr.2: 19481254 - 19484015 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02264276_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNE5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001036478.2 NP_001031555.1 3 - 4 408 93
NM_001084605.1 NP_001078074.1 3 - 4 408 93
NM_130315.5 NP_182269.1 3 - 4 574 93
NM_201979.2 NP_973708.1 3 - 4 408 93
GenBank mRNA AF083688.1 - 3 - 4 362 93
AJ271470.1 - 3 - 4 566 93
AK316899.2 - 3 - 4 394 93
AY074348.1 - 3 - 4 407 93
AY091388.1 - 3 - 4 325 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

embryo sac development
double fertilization forming a zygote and endosperm
embryo development ending in seed dormancy
response to endoplasmic reticulum stress
cell redox homeostasis
response to cadmium ion
pollen tube development
protein disulfide isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0