This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02263713_g1
See other SPA1 GE Assays ›
Gene Symbol:
SPA1
Gene Name:
SPA (suppressor of phyA-105) protein family
Gene Aliases:
AT2G46340, AT2G46350, F11C10.3, SUPPRESSOR OF PHYA-105 1
Chromosome Location:
Chr.2: 19022154 - 19027528 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02263713_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337186.1 NP_001325255.1 7 - 8 3495 79
NM_130197.5 NP_182157.2 7 - 8 3316 79
GenBank mRNA AF135455.1 - 7 - 8 3089 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
red, far-red light phototransduction
response to blue light
photomorphogenesis
chloroplast organization
red or far-red light signaling pathway
negative regulation of photomorphogenesis
response to red light
response to far red light
short-day photoperiodism, flowering
regulation of photoperiodism, flowering
protein kinase activity
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0