This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02263340_g1
See other SAP18 GE Assays ›
Gene Symbol:
SAP18
Gene Name:
SIN3 associated polypeptide P18
Gene Aliases:
AT2G45640, ATSAP18, F17K2.17, SIN3 ASSOCIATED POLYPEPTIDE 18, SIN3 associated polypeptide P18
Chromosome Location:
Chr.2: 18799528 - 18801471 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02263340_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SAP18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001202824.1 NP_001189753.1 1 - 2 260 110
NM_130126.4 NP_566050.1 1 - 2 172 110
GenBank mRNA AF378876.1 - 1 - 2 127 110
AK227453.1 - 1 - 2 123 110
AY050474.1 - 1 - 2 112 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

response to salt stress
response to abscisic acid
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0