This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02262903_g1
See other HUB1 GE Assays ›
Gene Symbol:
HUB1
Gene Name:
histone mono-ubiquitination 1
Gene Aliases:
AT2G44950, AT2G44960, RDO4, REDUCED DORMANCY 4, T14P1.25, T14P1_25, histone mono-ubiquitination 1
Chromosome Location:
Chr.2: 18542175 - 18548652 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02262903_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HUB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130060.4 NP_182022.2 17 - 18 2751 99
GenBank mRNA AY081322.1 - 17 - 18 2690 99
BT010360.1 - 17 - 18 2346 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein monoubiquitination
defense response to fungus, incompatible interaction
leaf morphogenesis
seed dormancy process
vegetative to reproductive phase transition of meristem
regulation of G2/M transition of mitotic cell cycle
histone monoubiquitination
histone H2B ubiquitination
innate immune response
regulation of timing of cell differentiation
cell division
positive regulation of cell division
ubiquitin-protein transferase activity
protein binding
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0