This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02261853_g1
See other CRTISO GE Assays ›
Gene Symbol:
CRTISO
Gene Name:
carotenoid isomerase
Gene Aliases:
AT1G06820, CAROTENOID AND CHLOROPLAST REGULATION 2, CAROTENOID ISOMERASE, CCR2, F4H5.10, F4H5_10, carotenoid isomerase
Chromosome Location:
Chr.1: 2092946 - 2096784 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02261853_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRTISO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331668.1 NP_001322208.1 8 - 9 1566 63
NM_001331669.1 NP_001322209.1 8 - 9 1566 63
NM_001331670.1 NP_001322210.1 8 - 9 1546 63
NM_100559.4 NP_172167.2 8 - 9 1805 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

etioplast organization
carotenoid biosynthetic process
oxidation-reduction process
oxidoreductase activity
carotenoid isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0