This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02258751_g1
See other AT1G01880 GE Assays ›
Gene Symbol:
AT1G01880
Gene Name:
5'-3' exonuclease family protein
Gene Aliases:
AT1G01880, AtGEN1, F22M8.2, F22M8_2, GEN1, ortholog of HsGEN1
Chromosome Location:
Chr.1: 306467 - 309541 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02258751_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G01880
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197958.1 NP_001184887.1 6 - 7 1679 134
NM_001331312.1 NP_001320987.1 8 - 9 1807 134
NM_001331313.1 NP_001320986.1 5 - 6 1398 134
NM_100069.3 NP_171691.2 6 - 7 1793 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

exodeoxyribonuclease

Gene Ontology Categories:

Function(s) Process(es)

DNA metabolic process
pollen development
UV protection
nucleic acid phosphodiester bond hydrolysis
DNA binding
crossover junction endodeoxyribonuclease activity
5'-flap endonuclease activity
flap endonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header