This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02255809_g1
See other NTL9 GE Assays ›
Gene Symbol:
NTL9
Gene Name:
NAC transcription factor-like 9
Gene Aliases:
AT4G35580, CBNAC, F8D20.90, F8D20_90, NAC transcription factor-like 9, calmodulin-binding NAC protein
Chromosome Location:
Chr.4: 16888386 - 16890877 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02255809_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NTL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001125650.1 NP_001119122.1 3 - 4 1077 69
NM_001203998.1 NP_001190927.1 3 - 4 1153 69
NM_119723.5 NP_567986.3 3 - 4 1151 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of defense response
negative regulation of transcription, DNA-templated
cellular response to osmotic stress
positive regulation of defense response to bacterium
transcription regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
calmodulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0