This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02255391_g1
See other AT3G20390 GE Assays ›
Gene Symbol:
AT3G20390
Gene Name:
endoribonuclease L-PSP family protein
Gene Aliases:
AT3G20390, Reactive Intermediate Deaminase A, RidA
Chromosome Location:
Chr.3: 7109862 - 7111929 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02255391_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G20390
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001338478.1 NP_001327365.1 2 - 3 535 114
NM_112930.3 NP_188674.1 2 - 3 438 114
GenBank mRNA AF375446.1 - 2 - 3 373 114
AK227774.1 - 2 - 3 406 114
AK229768.1 - 6 - 7 1393 114
AY060547.1 - 2 - 3 301 114
AY086036.1 - 2 - 3 430 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

deaminase

Gene Ontology Categories:

Function(s) Process(es)

branched-chain amino acid biosynthetic process
response to cadmium ion
organonitrogen compound catabolic process
deaminase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0