This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02254663_g1
See other PARC6 GE Assays ›
Gene Symbol:
PARC6
Gene Name:
plastid division protein
Gene Aliases:
A. THALIANA CHLOROPLAST DIVISION SITE POSITIONING 1, ARC6H, AT3G19180, ATCDP1, CDP1, CHLOROPLAST DIVISION SITE POSITIONING 1, paralog of ARC6
Chromosome Location:
Chr.3: 6632637 - 6637292 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02254663_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PARC6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035648.1 NP_001030725.1 9 - 10 1563 100
NM_112805.5 NP_188549.2 9 - 10 1602 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chloroplast fission
plastid fission
protein binding
protein self-association

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0