This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02250762_g1
See other AT1G80030 GE Assays ›
Gene Symbol:
AT1G80030
Gene Name:
Molecular chaperone Hsp40/DnaJ family protein
Gene Aliases:
AT1G80030, DJA7, DNA J protein A7, F18B13.12, F18B13_12
Chromosome Location:
Chr.1: 30104830 - 30109087 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02250762_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G80030
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001334948.1 NP_001321726.1 8 - 9 1350 86
NM_106652.3 NP_565227.1 8 - 9 1471 86
NM_179579.4 NP_849910.1 8 - 9 1471 86
NM_179580.2 NP_849911.1 8 - 9 1471 86
GenBank mRNA AK229599.1 - 8 - 9 1345 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to heat
protein refolding
chaperone mediated protein folding requiring cofactor
ATP binding
heat shock protein binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header