This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02249548_g1
See other AT4G33910 GE Assays ›
Gene Symbol:
AT4G33910
Gene Name:
2-oxoglutarate (2OG) and Fe(II)-dependent oxygenase superfamily protein
Gene Aliases:
AT4G33910, F17I5.100, F17I5_100
Chromosome Location:
Chr.4: 16256694 - 16258974 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02249548_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G33910
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119550.3 NP_567941.1 3 - 4 758 70
GenBank mRNA AY064069.1 - 3 - 4 543 70
AY086254.1 - 3 - 4 555 70
AY096377.1 - 3 - 4 398 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-proline hydroxylation to 4-hydroxy-L-proline
procollagen-proline 4-dioxygenase activity
iron ion binding
L-ascorbic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0