This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02247174_g1
See other SRF7 GE Assays ›
Gene Symbol:
SRF7
Gene Name:
STRUBBELIG-receptor family 7
Gene Aliases:
AT3G14350, STRUBBELIG-receptor family 7
Chromosome Location:
Chr.3: 4782757 - 4787243 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02247174_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRF7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_112293.5 NP_188052.2 2 - 3 465 83
NM_202582.1 NP_974311.1 1 - 2 521 83
NM_202583.1 NP_974312.1 2 - 3 389 83
GenBank mRNA AY518292.1 - 2 - 3 459 83
BT046202.1 - 2 - 3 221 83
FJ708721.1 - 2 - 3 221 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0