This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02245624_g1
See other DYL1 GE Assays ›
Gene Symbol:
DYL1
Gene Name:
dormancy-associated protein-like 1
Gene Aliases:
AT1G28330, AtDRM1, DORMANCY-ASSOCIATED PROTEIN 1, DRM1, F3H9.1, F3H9_1, dormancy-associated protein-like 1
Chromosome Location:
Chr.1: 9933786 - 9935440 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02245624_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DYL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_179389.4 NP_849720.1 2 - 3 387 82
NM_179390.1 NP_849721.1 2 - 3 385 82
GenBank mRNA AK317149.1 - 2 - 3 385 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to sucrose
response to glucose
response to fructose
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0