This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02245502_gH
See other PDIL1-1 GE Assays ›
Gene Symbol:
PDIL1-1
Gene Name:
PDI-like 1-1
Gene Aliases:
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 5, AT1G21750, ATPDI5, ATPDIL1-1, F8K7.19, F8K7_19, PDI-like 1-1, PDI5, PROTEIN DISULFIDE ISOMERASE 5
Chromosome Location:
Chr.1: 7645610 - 7648855 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02245502_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDIL1-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_102024.4 NP_173594.1 7 - 8 1341 81
NM_179365.1 NP_849696.1 7 - 8 1250 81
GenBank mRNA AY035096.1 - 7 - 8 1250 81
AY063059.1 - 7 - 8 1184 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
embryo development ending in seed dormancy
response to zinc ion
response to endoplasmic reticulum stress
regulation of programmed cell death
cell redox homeostasis
seed development
protein disulfide isomerase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0