This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02244044_g1
See other SVL1 GE Assays ›
Gene Symbol:
SVL1
Gene Name:
SHV3-like 1
Gene Aliases:
AT5G55480, GDPDL4, GPDL1, Glycerophosphodiester phosphodiesterase (GDPD) like 4, MTE17.19, MTE17_19, SHV3-like 1, glycerophosphodiesterase-like 1
Chromosome Location:
Chr.5: 22474142 - 22478216 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02244044_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SVL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124930.4 NP_200359.1 6 - 7 1503 99
GenBank mRNA BT015819.1 - 6 - 7 1415 99
BT020207.1 - 6 - 7 1368 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
trichome differentiation
guard cell morphogenesis
plant-type cell wall cellulose metabolic process
glycerophosphodiester phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0