This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02243832_g1
See other AT5G55220 GE Assays ›
Gene Symbol:
AT5G55220
Gene Name:
trigger factor type chaperone family protein
Gene Aliases:
AT5G55220, MCO15.17, MCO15_17
Chromosome Location:
Chr.5: 22397457 - 22400856 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02243832_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G55220
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124904.5 NP_200333.2 4 - 5 847 140
GenBank mRNA AY074845.1 - 4 - 5 688 140
BT002296.1 - 4 - 5 627 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein transport
protein unfolding
'de novo' cotranslational protein folding
chaperone-mediated protein folding
mRNA binding
peptidyl-prolyl cis-trans isomerase activity
ribosome binding
protein binding involved in protein folding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0