This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02243299_g1
See other ATM2 GE Assays ›
Gene Symbol:
ATM2
Gene Name:
myosin 2
Gene Aliases:
ARABIDOPSIS THALIANA MYOSIN 1, ARABIDOPSIS THALIANA MYOSIN 4, AT5G54280, ATM4, ATMYOS1, MDK4.10, MDK4_10, MYOSIN 4, myosin 2
Chromosome Location:
Chr.5: 22039383 - 22046358 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02243299_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085286.2 NP_001078755.1 13 - 14 2123 110
NM_124808.4 NP_568806.3 12 - 13 1247 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
actin filament-based movement
vesicle transport along actin filament
microfilament motor activity
motor activity
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm