This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02242112_g1
See other EMB2757 GE Assays ›
Gene Symbol:
EMB2757
Gene Name:
Transducin/WD40 repeat-like superfamily protein
Gene Aliases:
ALT2, ALUMINUM TOLERANT 2, AT4G29860, AtDSE1, DECREASED SIZE EXCLUSION LIMIT 1, DSE1, EMBRYO DEFECTIVE 2757, F27B13.100, F27B13_100, TAN, TANMEI
Chromosome Location:
Chr.4: 14601698 - 14605810 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02242112_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMB2757
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119132.3 NP_194715.2 6 - 7 649 107
GenBank mRNA AB191306.1 - 6 - 7 625 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

regulation of DNA repair
cellular response to DNA damage stimulus
pollen development
plasmodesma organization
embryo development ending in seed dormancy
response to aluminum ion
plasmodesmata-mediated intercellular transport
DNA integrity checkpoint
plant ovule development
acquisition of desiccation tolerance in seed
positive regulation of cell cycle arrest
response to cisplatin
regulation of root development

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0